8-(3,5-Dimethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

130010-1G 130010-500MG 130010-5G 130010-25G
77464-596EA 397.58 USD
77464-596 77477-382 77487-484 77498-272
8-(3,5-Dimethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(3,5-Dimethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₆H₂₂O₃
Storage Temperature: Ambient
MDL Number: MFCD21594356
CAS Number: 1466024-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR