8-(4-(dimethylamino)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129988-1G 129988-500MG 129988-5G 129988-25G
77464-588EA 415.09 USD
77464-588 77477-374 77487-476 77498-264
8-(4-(dimethylamino)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(4-(dimethylamino)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₆H₂₃NO₃
MW: 277.359 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32186354
CAS Number: 117960-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR