8-(2-(dimethylamino)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129984-1G 129984-500MG 129984-5G 129984-25G
77464-584EA 415.09 USD
77464-584 77477-370 77487-472 77498-260
8-(2-(dimethylamino)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(2-(dimethylamino)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₆H₂₃NO₃
MW: 277.359 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34828279
CAS Number: 2921756-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR