8-(2-(trifluoromethyl)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

129976-1G 129976-500MG 129976-5G 129976-25G
77464-580EA 415.09 USD
77464-580 77477-366 77487-468 77498-256
8-(2-(trifluoromethyl)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(2-(trifluoromethyl)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₅H₁₇F₃O₃
MW: 302.289 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34828278
CAS Number: 1472679-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR