8-(4-Ethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129966-1G 129966-500MG 129966-5G 129966-25G
77464-576EA 397.58 USD
77464-576 77477-362 77487-464 77498-252
8-(4-Ethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(4-Ethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₆H₂₂O₃
Storage Temperature: Ambient
MDL Number: MFCD20862266
CAS Number: 1477712-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR