8-(3-Ethoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

129952-1G 129952-500MG 129952-5G
77464-568EA 497.49 USD
77464-568 77477-354 77487-456
8-(3-Ethoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(3-Ethoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₆H₂₂O₄
MW: 278.343 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26743336
CAS Number: 1508547-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR