8-(4-Methoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129948-1G 129948-500MG 129948-5G
77464-564EA 459.38 USD
77464-564 77477-350 77487-452
8-(4-Methoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(4-Methoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₅H₂₀O₄
Storage Temperature: Ambient
MDL Number: MFCD21078742
CAS Number: 67019-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR