8-(p-tolyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129942-1G 129942-500MG 129942-5G 129942-25G
77464-560EA 397.58 USD
77464-560 77477-346 77487-448 77498-242
8-(p-tolyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(p-tolyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₅H₂₀O₃
Storage Temperature: Ambient
MDL Number: MFCD21594336
CAS Number: 365553-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR