1-(2-Fluoro-5-methylphenyl)cycloheptanol ≥95%

Supplier: AOBChem USA

Warning

129826-1G 129826-500MG 129826-5G 129826-25G
77464-556EA 415.09 USD
77464-556 77477-342 77487-444 77498-238
1-(2-Fluoro-5-methylphenyl)cycloheptanol ≥95%
1-(2-Fluoro-5-methylphenyl)cycloheptanol

Precaución: For research use only

Formula: C₁₄H₁₉FO
MW: 222.2984 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222262
CAS Number: 1343691-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR