3-Hydroxy-2-methyl-3-(4-(trifluoromethyl)phenyl)isoindolin-1-one ≥97%

Supplier: AOBChem USA

Warning

129624-1G 129624-500MG 129624-5G 129624-25G
77464-552EA 415.09 USD
77464-552 77477-338 77487-440 77498-234
3-Hydroxy-2-methyl-3-(4-(trifluoromethyl)phenyl)isoindolin-1-one ≥97%
3-Hydroxy-2-methyl-3-(4-(trifluoromethyl)phenyl)isoindolin-1-one

Precaución: For research use only

Formula: C₁₆H₁₂F₃NO₂
MW: 307.267 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34828171
CAS Number: 153397-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR