3-(4-Bromophenyl)-3-hydroxy-2-methylisoindolin-1-one ≥95%

Supplier: AOBChem USA

Warning

129618-1G 129618-500MG 129618-5G 129618-25G
77464-548EA 415.09 USD
77464-548 77477-334 77487-436 77498-230
3-(4-Bromophenyl)-3-hydroxy-2-methylisoindolin-1-one ≥95%
3-(4-Bromophenyl)-3-hydroxy-2-methylisoindolin-1-one

Precaución: For research use only

Formula: C₁₅H₁₂BrNO₂
MW: 318.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34828166
CAS Number: 2969339-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR