1-(3-Chloro-4-methylphenyl)cyclooctanol ≥95%

Supplier: AOBChem USA

Warning

129570-1G 129570-500MG 129570-5G 129570-25G
77464-544EA 415.09 USD
77464-544 77477-330 77487-432 77498-226
1-(3-Chloro-4-methylphenyl)cyclooctanol ≥95%
1-(3-Chloro-4-methylphenyl)cyclooctanol

Precaución: For research use only

Formula: C₁₅H₂₁ClO
MW: 252.78 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34828158
CAS Number: 2969370-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR