1-(3,5-Dichlorophenyl)cyclooctanol ≥95%

Supplier: AOBChem USA

Warning

129561-1G 129561-500MG 129561-5G 129561-25G
77464-540EA 415.09 USD
77464-540 77477-326 77487-428 77498-222
1-(3,5-Dichlorophenyl)cyclooctanol ≥95%
1-(3,5-Dichlorophenyl)cyclooctanol

Precaución: For research use only

Formula: C₁₄H₁₈Cl₂O
MW: 273.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34828155
CAS Number: 2921687-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR