1-(4-Fluorophenyl)cyclooctanol ≥97%

Supplier: AOBChem USA

Warning

129531-1G 129531-500MG 129531-5G 129531-25G
77464-532EA 415.09 USD
77464-532 77477-318 77487-420 77498-214
1-(4-Fluorophenyl)cyclooctanol ≥97%
1-(4-Fluorophenyl)cyclooctanol

Precaución: For research use only

Formula: C₁₄H₁₉FO
MW: 222.298 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34828145
CAS Number: 908095-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR