6-Bromo-5-fluoro-3-iodo-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

129343-1G 129343-500MG 129343-5G
77464-524EA 441.87 USD
77464-524 77477-310 77487-412
6-Bromo-5-fluoro-3-iodo-1H-indazole ≥95%
6-Bromo-5-fluoro-3-iodo-1H-indazole

Precaución: For research use only

Formula: C₇H₃BrFIN₂
MW: 340.919 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384871
CAS Number: 1286734-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR