3-Fluoro-2-(trifluoromethyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (3-Fluoro-2-(trifluoromethyl)phenyl)boronic acid, 2-(Trifluoromethyl)-3-fluoro-phenylboronic acid

12915-1G 12915-500MG 12915-5G 12915-25G
77464-520EA 176.13 USD
77464-520 77477-306 77487-408 77498-204
3-Fluoro-2-(trifluoromethyl)phenylboronic acid ≥95%
3-Fluoro-2-(trifluoromethyl)phenylboronic acid

Precaución: For research use only

Formula: C₇H₅BF₄O₂
MW: 207.918 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08706086
CAS Number: 850411-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR