(2,6-Difluorophenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

128325-1G 128325-500MG 128325-5G 128325-25G
77464-516EA 347.11 USD
77464-516 77477-302 77487-404 77498-200
(2,6-Difluorophenyl)(furan-2-yl)methanol ≥95%
(2,6-Difluorophenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD19612735
CAS Number: 1339584-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR