(3-Chloro-4-methylphenyl)(furan-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

128319-1G 128319-500MG 128319-5G 128319-25G
77464-512EA 397.58 USD
77464-512 77477-298 77487-400 77498-196
(3-Chloro-4-methylphenyl)(furan-2-yl)methanol ≥97%
(3-Chloro-4-methylphenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₂H₁₁ClO₂
MW: 222.668 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34827573
CAS Number: 1592394-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR