(2,4-Difluorophenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

128309-1G 128309-500MG 128309-5G 128309-25G
77464-508EA 297.67 USD
77464-508 77477-294 77487-396 77498-192
(2,4-Difluorophenyl)(furan-2-yl)methanol ≥95%
(2,4-Difluorophenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₈F₂O₂
MW: 210.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787326
CAS Number: 1250225-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR