1-(3-Chloro-4-methylphenyl)cycloheptanol ≥95%

Supplier: AOBChem USA

Warning

128277-1G 128277-500MG 128277-5G 128277-25G
77464-504EA 415.09 USD
77464-504 77477-290 77487-392 77498-188
1-(3-Chloro-4-methylphenyl)cycloheptanol ≥95%
1-(3-Chloro-4-methylphenyl)cycloheptanol

Precaución: For research use only

Formula: C₁₄H₁₉ClO
MW: 238.753 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34827569
CAS Number: 1693617-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR