1-(4-(trifluoromethoxy)phenyl)cycloheptanol ≥95%

Supplier: AOBChem USA

Warning

128271-1G 128271-500MG 128271-5G 128271-25G
77464-500EA 415.09 USD
77464-500 77477-286 77487-388 77498-184
1-(4-(trifluoromethoxy)phenyl)cycloheptanol ≥95%
1-(4-(trifluoromethoxy)phenyl)cycloheptanol

Precaución: For research use only

Formula: C₁₄H₁₇F₃O₂
MW: 274.279 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34827567
CAS Number: 2969358-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR