1-(4-(trifluoromethyl)phenyl)cycloheptanol ≥95%

Supplier: AOBChem USA

Warning

128256-1G 128256-500MG 128256-5G 128256-25G
77464-496EA 415.09 USD
77464-496 77477-282 77487-384 77498-180
1-(4-(trifluoromethyl)phenyl)cycloheptanol ≥95%
1-(4-(trifluoromethyl)phenyl)cycloheptanol

Precaución: For research use only

Formula: C₁₄H₁₇F₃O
MW: 258.279 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18022805
CAS Number: 77826-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR