1-(3-Fluorophenyl)cycloheptanol ≥95%

Supplier: AOBChem USA

Warning

128244-1G 128244-500MG 128244-5G 128244-25G
77464-492EA 415.09 USD
77464-492 77477-278 77487-380 77498-176
1-(3-Fluorophenyl)cycloheptanol ≥95%
1-(3-Fluorophenyl)cycloheptanol

Precaución: For research use only

Formula: C₁₃H₁₇FO
MW: 208.272 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12152940
CAS Number: 77826-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR