1-(5-Bromo-2-fluorophenyl)cycloheptanol ≥95%

Supplier: AOBChem USA

Warning

128238-1G 128238-500MG 128238-5G
77464-488EA 447.02 USD
77464-488 77477-274 77487-376
1-(5-Bromo-2-fluorophenyl)cycloheptanol ≥95%
1-(5-Bromo-2-fluorophenyl)cycloheptanol

Precaución: For research use only

Formula: C₁₃H₁₆BrFO
MW: 287.168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23759087
CAS Number: 1508670-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR