3-Bromo-2-iodo-4-methylthiophene ≥95%

Supplier: AOBChem USA

Warning

127954-1G 127954-500MG 127954-5G 127954-25G
77464-484EA 415.09 USD
77464-484 77477-270 77487-370 77498-168
3-Bromo-2-iodo-4-methylthiophene ≥95%
3-Bromo-2-iodo-4-methylthiophene

Precaución: For research use only

Formula: C₅H₄BrIS
Storage Temperature: Ambient
MDL Number: MFCD34827401
CAS Number: 2921910-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR