3-Bromo-4-methylthiophene-2-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

127950-1G 127950-500MG 127950-5G 127950-25G
77464-480EA 329.6 USD
77464-480 77477-266 77487-366 77498-164
3-Bromo-4-methylthiophene-2-carboxylic acid ≥95%
3-Bromo-4-methylthiophene-2-carboxylic acid

Precaución: For research use only

Formula: C₆H₅BrO₂S
MW: 221.072 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426927
CAS Number: 854627-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR