Methyl 2,4,6-trichloronicotinate ≥95%

Supplier: AOBChem USA

Warning

127707-1G 127707-500MG 127707-5G 127707-25G
77464-476EA 168.92 USD
77464-476 77477-262 77487-362 77498-160
Methyl 2,4,6-trichloronicotinate ≥95%
Methyl 2,4,6-trichloronicotinate

Precaución: For research use only

Formula: C₇H₄Cl₃NO₂
MW: 240.471 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21604740
CAS Number: 1218994-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR