3,4-Dichloro-5-hydroxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

127090-1G 127090-500MG 127090-5G
77464-472EA 723.06 USD
77464-472 77477-258 77487-358
3,4-Dichloro-5-hydroxybenzoic acid ≥95%
3,4-Dichloro-5-hydroxybenzoic acid

Precaución: For research use only

Formula: C₇H₄Cl₂O₃
MW: 207.011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24480406
CAS Number: 63001-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR