1-(1-Fluoronaphthalen-2-yl)ethanol ≥97%

Supplier: AOBChem USA

Warning

127031-1G 127031-5G 127031-25G
77464-468EA 60.77 USD
77464-468 77487-352 77498-154
1-(1-Fluoronaphthalen-2-yl)ethanol ≥97%
1-(1-Fluoronaphthalen-2-yl)ethanol

Precaución: For research use only

Formula: C₁₂H₁₁FO
MW: 190.214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34826815
CAS Number: 1880380-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR