2-Bromo-4-fluoro-3-methylbenzonitrile ≥95%

Supplier: AOBChem USA

Warning

127013-1G 127013-500MG 127013-5G
77464-464EA 1065.02 USD
77464-464 77477-252 77487-348
2-Bromo-4-fluoro-3-methylbenzonitrile ≥95%
2-Bromo-4-fluoro-3-methylbenzonitrile

Precaución: For research use only

Formula: C₈H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD28739734
CAS Number: 1805592-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR