4'-Cyano-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

126827-1G 126827-500MG 126827-5G 126827-25G
77464-460EA 182.31 USD
77464-460 77477-248 77487-340 77498-144
4'-Cyano-2,2,2-trifluoroacetophenone ≥97%
4'-Cyano-2,2,2-trifluoroacetophenone

Precaución: For research use only

Formula: C₉H₄F₃NO
Storage Temperature: Ambient
MDL Number: MFCD01319971
CAS Number: 23516-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR