1,2-Difluoro-3-iodo-4-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

126818-1G 126818-500MG 126818-5G 126818-25G
77464-456EA 158.62 USD
77464-456 77477-244 77487-336 77498-140
1,2-Difluoro-3-iodo-4-(methoxymethoxy)benzene ≥97%
1,2-Difluoro-3-iodo-4-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₇F₂IO₂
MW: 300.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34826716
CAS Number: 2918964-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR