1-(5-Bromo-2-fluorophenyl)cyclobutanol ≥97%

Supplier: AOBChem USA

Warning

126500-1G 126500-500MG 126500-5G 126500-25G
77464-452EA 340.93 USD
77464-452 77477-240 77487-332 77498-136
1-(5-Bromo-2-fluorophenyl)cyclobutanol ≥97%
1-(5-Bromo-2-fluorophenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₀H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD23759131
CAS Number: 1516561-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR