1-(6-Bromo-2-fluoro-3-methylphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

126371-1G 126371-500MG 126371-5G 126371-25G
77464-448EA 415.09 USD
77464-448 77477-236 77487-328 77498-132
1-(6-Bromo-2-fluoro-3-methylphenyl)propan-1-ol ≥95%
1-(6-Bromo-2-fluoro-3-methylphenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₂BrFO
MW: 247.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34826517
CAS Number: 2969310-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR