1-(5-Bromothiophen-2-yl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

125591-1G 125591-500MG 125591-5G 125591-25G
77464-444EA 297.67 USD
77464-444 77477-232 77487-324 77498-124
1-(5-Bromothiophen-2-yl)-2-methylpropan-1-ol ≥95%
1-(5-Bromothiophen-2-yl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₈H₁₁BrOS
MW: 235.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12788261
CAS Number: 1216221-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR