1-(5-Bromothiophen-2-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

125585-1G 125585-500MG 125585-5G
77464-440EA 572.68 USD
77464-440 77477-228 77487-320
1-(5-Bromothiophen-2-yl)-2,2-difluoroethanone ≥95%
1-(5-Bromothiophen-2-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₆H₃BrF₂OS
MW: 241.053 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28106457
CAS Number: 1314893-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR