phenyl(thiophen-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

125573-1G 125573-500MG 125573-5G 125573-25G
77464-436EA 229.69 USD
77464-436 77477-224 77487-314 77498-118
phenyl(thiophen-2-yl)methanol ≥97%
phenyl(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₁₀OS
MW: 190.262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00277660
CAS Number: 26059-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR