tert-Butyl 4-(furan-2-yl)-4-hydroxypiperidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

125558-1G 125558-500MG 125558-5G
77464-432EA 522.21 USD
77464-432 77477-220 77487-310
tert-Butyl 4-(furan-2-yl)-4-hydroxypiperidine-1-carboxylate ≥95%
tert-Butyl 4-(furan-2-yl)-4-hydroxypiperidine-1-carboxylate

Precaución: For research use only

Formula: C₁₄H₂₁NO₄
MW: 267.321 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26711411
CAS Number: 1823257-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR