5-Fluorobenzofuran-7-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

125543-1G 125543-500MG 125543-5G
77464-428EA 644.78 USD
77464-428 77477-214 77487-304
5-Fluorobenzofuran-7-carboxylic acid ≥97%
5-Fluorobenzofuran-7-carboxylic acid

Precaución: For research use only

Formula: C₉H₅FO₃
MW: 180.1326 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22560160
CAS Number: 1388021-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR