3-tert-Butyl-5-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-t-Butyl-5-methylphenylboronic acid

12554-1G 12554-500MG 12554-5G 12554-25G
77464-424EA 120.51 USD
77464-424 77477-216 77487-306 77498-114
3-tert-Butyl-5-methylphenylboronic acid ≥97%
3-tert-Butyl-5-methylphenylboronic acid

Precaución: For research use only

Formula: C₁₁H₁₇BO₂
Storage Temperature: Ambient
MDL Number: MFCD03411938
CAS Number: 193905-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR