tert-Butyl 4-(2-fluoro-5-methoxyphenyl)-4-hydroxypiperidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

125491-1G 125491-500MG 125491-5G 125491-25G
77464-420EA 415.09 USD
77464-420 77477-208 77487-300 77498-112
tert-Butyl 4-(2-fluoro-5-methoxyphenyl)-4-hydroxypiperidine-1-carboxylate ≥95%
tert-Butyl 4-(2-fluoro-5-methoxyphenyl)-4-hydroxypiperidine-1-carboxylate

Precaución: For research use only

Formula: C₁₇H₂₄FNO₄
Storage Temperature: Ambient
MDL Number: MFCD34826189
CAS Number: 2969310-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR