8-(3-Bromo-2,6-difluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

125474-1G 125474-500MG 125474-5G 125474-25G
77464-416EA 415.09 USD
77464-416 77477-204 77487-296 77498-108
8-(3-Bromo-2,6-difluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(3-Bromo-2,6-difluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₄H₁₅BrF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34826182
CAS Number: 1969534-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR