4-(4-Fluorophenyl)-4-hydroxycyclohexanone ≥97%

Supplier: AOBChem USA

Warning

125446-1G 125446-500MG 125446-5G
77464-412EA 574.74 USD
77464-412 77477-200 77487-292
4-(4-Fluorophenyl)-4-hydroxycyclohexanone ≥97%
4-(4-Fluorophenyl)-4-hydroxycyclohexanone

Precaución: For research use only

Formula: C₁₂H₁₃FO₂
MW: 208.2288 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21321193
CAS Number: 36716-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR