3-Bromo-2-(cyclopentyloxy)-5-methylpyridine ≥96%

Supplier: AOBChem USA

Warning

125411-1G 125411-500MG 125411-5G 125411-25G
77464-408EA 347.11 USD
77464-408 77477-196 77487-288 77498-102
3-Bromo-2-(cyclopentyloxy)-5-methylpyridine ≥96%
3-Bromo-2-(cyclopentyloxy)-5-methylpyridine

Precaución: For research use only

Formula: C₁₁H₁₄BrNO
MW: 256.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29114184
CAS Number: 1289007-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR