4-((benzyloxy)methyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

125231-1G 125231-500MG 125231-5G 125231-25G
77464-404EA 210.12 USD
77464-404 77477-192 77487-284 77498-098
4-((benzyloxy)methyl)benzoic acid ≥95%
4-((benzyloxy)methyl)benzoic acid

Precaución: For research use only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Ambient
CAS Number: 22714-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR