2-Bromo-3,4-dichloropyridine ≥97%

Supplier: AOBChem USA

Warning

125015-1G 125015-500MG 125015-5G 125015-25G
77464-400EA 146.26 USD
77464-400 77477-188 77487-280 77498-092
2-Bromo-3,4-dichloropyridine ≥97%
2-Bromo-3,4-dichloropyridine

Precaución: For research use only

Formula: C₅H₂BrCl₂N
Storage Temperature: Ambient
MDL Number: MFCD28737853
CAS Number: 1807009-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR