4-(5-Chloropyridin-3-yl)-1-methylpiperidin-4-ol ≥97%

Supplier: AOBChem USA

Warning

124837-1G 124837-500MG 124837-5G 124837-25G
77464-396EA 415.09 USD
77464-396 77477-184 77487-276 77498-088
4-(5-Chloropyridin-3-yl)-1-methylpiperidin-4-ol ≥97%
4-(5-Chloropyridin-3-yl)-1-methylpiperidin-4-ol

Precaución: For research use only

Formula: C₁₁H₁₅ClN₂O
MW: 226.7026 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34825944
CAS Number: 2969337-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR