tert-Butyl 4-(3,4-dichlorophenyl)-4-hydroxypiperidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

124783-1G 124783-500MG 124783-5G 124783-25G
77464-392EA 254.41 USD
77464-392 77477-180 77487-272 77498-084
tert-Butyl 4-(3,4-dichlorophenyl)-4-hydroxypiperidine-1-carboxylate ≥95%
tert-Butyl 4-(3,4-dichlorophenyl)-4-hydroxypiperidine-1-carboxylate

Precaución: For research use only

Formula: C₁₆H₂₁Cl₂NO₃
MW: 346.2488 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11849845
CAS Number: 256958-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR