tert-Butyl 4-(3-bromophenyl)-4-hydroxypiperidine-1-carboxylate ≥95%
Supplier: AOBChem USA
|
Warning
|
124771-1G
124771-500MG
124771-5G
77464-388EA
457.32
USD
77464-388
77477-176
77487-268
tert-Butyl 4-(3-bromophenyl)-4-hydroxypiperidine-1-carboxylate ≥95%
tert-Butyl 4-(3-bromophenyl)-4-hydroxypiperidine-1-carboxylate
Precaución: For research use only
Formula:
C₁₆H₂₂BrNO₃ MW: 356.2547 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09953414 CAS Number: 1198286-72-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...