2-Chloro-1-(2,5-dibromopyridin-4-yl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

124619-1G 124619-500MG 124619-5G 124619-25G
77464-376EA 415.09 USD
77464-376 77477-164 77487-256 77498-076
2-Chloro-1-(2,5-dibromopyridin-4-yl)-2,2-difluoroethanone ≥97%
2-Chloro-1-(2,5-dibromopyridin-4-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₇H₂Br₂ClF₂NO
MW: 349.355 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34825911
CAS Number: 2969309-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR